Anti-BID Goat polyclonal antibody

Lieferant: OriGene
ORIGTA302648EA 539 EUR
ORIGTA302648
Anti-BID Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:8000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR