Anti-APEX1 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302643EA 536 EUR
ORIGTA302643
Anti-APEX1 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1-0.3?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR