Anti-YTHDF3 Rabbit Polyclonal Antibody [clone: 403CT8.5.5]

Lieferant: Abgent
ABGEAP9985AEA 350 EUR
ABGEAP9985A
Anti-YTHDF3 Rabbit Polyclonal Antibody [clone: 403CT8.5.5]
Antikörper
Type: Primary
Antigen: YTHDF3 (YTH domain family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR