Anti-YPEL5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9958BEA 348 EUR
ABGEAP9958B
Anti-YPEL5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: YPEL5 (yippee-like 5 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR