Anti-SDPR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9935CEA 348 EUR
ABGEAP9935C
Anti-SDPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SDPR (serum deprivation response)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR