Anti-NEDD4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9934BEA 350 EUR
ABGEAP9934B
Anti-NEDD4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NEDD4 (neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR