Anti-B3GNT6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9886CEA 350 EUR
ABGEAP9886C
Anti-B3GNT6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B3GNT6 (UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR