Anti-BSND Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9858BEA 350 EUR
ABGEAP9858B
Anti-BSND Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BSND (Bartter syndrome, infantile, with sensorineural deafness (Barttin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR