Anti-NDUFA10 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9768CEA 348 EUR
ABGEAP9768C
Anti-NDUFA10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NDUFA10 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR