Anti-GDA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9710AEA 348 EUR
ABGEAP9710A
Anti-GDA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GDA (guanine deaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR