Anti-GGCT Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9676BEA 348 EUR
ABGEAP9676B
Anti-GGCT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GGCT (gamma-glutamylcyclotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR