Anti-BCL11A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9630BEA 350 EUR
ABGEAP9630B
Anti-BCL11A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCL11A (B-cell CLL/lymphoma 11A (zinc finger protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR