Anti-ABCD2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9627CEA 350 EUR
ABGEAP9627C
Anti-ABCD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABCD2 (ATP-binding cassette, sub-family D (ALD), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR