Anti-CLPB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9613BEA 350 EUR
ABGEAP9613B
Anti-CLPB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLPB (ClpB caseinolytic peptidase B homolog (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR