Anti-GNAZ Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9608AEA 348 EUR
ABGEAP9608A
Anti-GNAZ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNAZ (guanine nucleotide binding protein (G protein), alpha z polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR