Anti-TMEM108 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9598BEA 350 EUR
ABGEAP9598B
Anti-TMEM108 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM108 (transmembrane protein 108)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR