Anti-SGIP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9556BEA 348 EUR
ABGEAP9556B
Anti-SGIP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SGIP1 (SH3-domain GRB2-like (endophilin) interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR