Anti-GINS1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9494AEA 360 EUR
ABGEAP9494A
Anti-GINS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GINS1 (GINS complex subunit 1 (Psf1 homolog))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR