Anti-NPY2R Rabbit Polyclonal Antibody [clone: 364CT25.4.2]

Lieferant: Abgent
ABGEAP9466AEA 350 EUR
ABGEAP9466A
Anti-NPY2R Rabbit Polyclonal Antibody [clone: 364CT25.4.2]
Antikörper
Type: Primary
Antigen: NPY2R (neuropeptide Y receptor Y2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR