Anti-COQ5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9319CEA 348 EUR
ABGEAP9319C
Anti-COQ5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COQ5 (coenzyme Q5 homolog, methyltransferase (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR