Anti-CDKN1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9311AEA 360 EUR
ABGEAP9311A
Anti-CDKN1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDKN1A (cyclin-dependent kinase inhibitor 1A (p21, Cip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR