Anti-DSTN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9189CEA 350 EUR
ABGEAP9189C
Anti-DSTN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DSTN (destrin (actin depolymerizing factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR