Anti-RPL18A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9141BEA 350 EUR
ABGEAP9141B
Anti-RPL18A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPL18A (ribosomal protein L18a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR