Anti-EFEMP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9095AEA 348 EUR
ABGEAP9095A
Anti-EFEMP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EFEMP1 (EGF containing fibulin-like extracellular matrix protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR