Anti-ITGA2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9051BEA 350 EUR
ABGEAP9051B
Anti-ITGA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGA2 (integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR