Anti-KIR2DL4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9042BEA 348 EUR
ABGEAP9042B
Anti-KIR2DL4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIR2DL4 (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR