Anti-DAB1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8986CEA 348 EUR
ABGEAP8986C
Anti-DAB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DAB1 (Dab, reelin signal transducer, homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR