Anti-RAB8A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8962AEA 348 EUR
ABGEAP8962A
Anti-RAB8A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAB8A (RAB8A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR