Anti-ACER3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8953BEA 350 EUR
ABGEAP8953B
Anti-ACER3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACER3 (alkaline ceramidase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR