Anti-CLIP1 Rabbit Polyclonal Antibody [clone: 363CT4.1.6]

Lieferant: Abgent
ABGEAP8950AEA 350 EUR
ABGEAP8950A
Anti-CLIP1 Rabbit Polyclonal Antibody [clone: 363CT4.1.6]
Antikörper
Type: Primary
Antigen: CLIP1 (CAP-GLY domain containing linker protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR