Anti-TRIB2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8930AEA 350 EUR
ABGEAP8930A
Anti-TRIB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIB2 (tribbles pseudokinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR