Anti-ABHD12 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8904AEA 350 EUR
ABGEAP8904A
Anti-ABHD12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABHD12 (abhydrolase domain containing 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR