Anti-QDPR Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8841BEA 350 EUR
ABGEAP8841B
Anti-QDPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: QDPR (quinoid dihydropteridine reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR