Anti-SLC3A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8802CEA 348 EUR
ABGEAP8802C
Anti-SLC3A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC3A1 (solute carrier family 3 (amino acid transporter heavy chain), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR