Anti-XPA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8784BEA 350 EUR
ABGEAP8784B
Anti-XPA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XPA (xeroderma pigmentosum, complementation group A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR