Anti-CDC42EP3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8775AEA 348 EUR
ABGEAP8775A
Anti-CDC42EP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDC42EP3 (CDC42 effector protein (Rho GTPase binding) 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR