Anti-CYP2C18 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8725BEA 350 EUR
ABGEAP8725B
Anti-CYP2C18 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP2C18 (cytochrome P450, family 2, subfamily C, polypeptide 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR