Anti-ALKBH3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8682BEA 350 EUR
ABGEAP8682B
Anti-ALKBH3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALKBH3 (alkB, alkylation repair homolog 3 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR