Anti-KLC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8637CEA 370 EUR
ABGEAP8637C
Anti-KLC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLC1 (kinesin light chain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR