Anti-ESRRA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8635CEA 350 EUR
ABGEAP8635C
Anti-ESRRA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ESRRA (estrogen-related receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR