Anti-ABCC5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8578CEA 350 EUR
ABGEAP8578C
Anti-ABCC5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ABCC5 (ATP-binding cassette, sub-family C (CFTR/MRP), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR