Anti-CDC14A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8440AEA 348 EUR
ABGEAP8440A
Anti-CDC14A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDC14A (cell division cycle 14A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR