Anti-PTEN Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8436AEA 360 EUR
ABGEAP8436A
Anti-PTEN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTEN (phosphatase and tensin homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR