Anti-PTPN3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8426AEA 348 EUR
ABGEAP8426A
Anti-PTPN3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPN3 (protein tyrosine phosphatase, non-receptor type 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR