Anti-MLST8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8200CEA 350 EUR
ABGEAP8200C
Anti-MLST8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR