Anti-PIK3CG Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8180EEA 348 EUR
ABGEAP8180E
Anti-PIK3CG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIK3CG (phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR