Anti-PLAU Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8161AEA 360 EUR
ABGEAP8161A
Anti-PLAU Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLAU (plasminogen activator, urokinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR