Anti-AK1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8160BEA 350 EUR
ABGEAP8160B
Anti-AK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AK1 (adenylate kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR