Anti-YWHAZ Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8152DEA 350 EUR
ABGEAP8152D
Anti-YWHAZ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: YWHAZ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR