Anti-PFKFB1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8147CEA 348 EUR
ABGEAP8147C
Anti-PFKFB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PFKFB1 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR